Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Polycystin-1, Clone: E4 (5F4D2),

Catalog No. MABS1252 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1252 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1252 Supplier MilliporeSigma™ Supplier No. MABS1252
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Polycystin-1 clone: E4 (5F4D2) in Human, Mouse, Rat samples, and it is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting

Polycystin-1 (UniProt P98161; also known as Autosomal dominant polycystic kidney disease 1 protein) is encoded by the PKD1 gene (Gene ID 5310) in human. The primary cilium is a microtubule-based mechano- and chemo-sensory organelle that coordinates an array of cellular pathways during development and in tissue homeostasis, including Hedgehog (Hh), PDGFRα and Wnt signaling. Defective ciliary function causes a variety of ciliopathies, including autosomal dominant polycystic kidney disease (ADPKD). Polycystin-1 (PC1) and polycystin-2 (PC2) are large transmembrane proteins co-localized to primary cilium, where they play an important role in calcium-based signalling. PC1 is an atypical adhesion G-protein-coupled receptor (aGPCR) with 11-transmemrbane domains. PC1 is cis-autoproteolytically cleaved at a juxtamembrane GPCR cleavage site (GPS; a.a. 3012-3061) in a developmental stage-dependent manner, where PC1 is largely uncleaved in early embryonic kidneys but becomes extensively cleaved after birth. GPS cleavage results in a heterodimeric PC1 form, in which the N-terminal fragment (NTF) remains non-covalently associated with the transmembrane C-terminal fragment (CTF). Transgenic mice expressing non-cleavable PC1 develop cystic kidney disease during the postnatal period.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Polycystin-1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone E4 (5F4D2)
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P98161
Gene Symbols PKD1
Host Species Mouse
Immunogen FLAG-tagged recombinant human polycystin-1 C-type lectin domain.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001009944.2
Target Species Human, Mouse, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.