Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polyoma virus, Large T, Mouse, Clone: PyLT, Abnova™
Mouse monoclonal antibody raised against polyoma virus, Large T antigen.
Supplier: Abnova Corporation MAB7375
Specifications
Polyoma virus, Large T | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Mouse | |
100 μg | |
Primary | |
Virus | |
Liquid |
Immunofluorescence, Immunoprecipitation, Western Blot | |
PyLT | |
Mouse monoclonal antibody raised against polyoma virus, Large T antigen. | |
In PBS | |
Polyoma virus-transformed Wistar rat fibroblast cell line Py REWA5/T1A1. | |
RUO | |
Polyoma virus, Large T antigen (PyLT). WB: Detects a band of approximately 100 KDa. | |
Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction