Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POP7, Mouse, Clone: 5BH5, Abnova™

Catalog No. 89112214 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-214 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-214 Supplier Abnova Corporation Supplier No. MAB5302

Mouse monoclonal antibody raised against recombinant POP7.

Specifications

Antigen POP7
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5BH5
Conjugate Unconjugated
Description processing of precursor 7, ribonuclease P/MRP subunit (S. cerevisiae)
Formulation Lyophilized from 10mM ammonium bicarbonate buffer (2mg BSA)
Gene POP7
Gene Alias 0610037N12Rik, RPP2, RPP20
Gene Symbols POP7
Host Species Mouse
Immunogen Recombinant GST fusion protein corresponding to human POP7.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10248
Target Species Human
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.