Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPIB, Mouse, Clone: k2E2, Abnova™

Catalog No. 89110775 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-775 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-775 Supplier Abnova Corporation Supplier No. MAB2033
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant PPIB.

The protein encoded by this gene is a cyclosporine-binding protein and is mainly located within the endoplasmic reticulum. It is associated with the secretory pathway and released in biological fluids. This protein can bind to cells derived from T- and B-lymphocytes, and may regulate cyclosporine A-mediated immunosuppression. [provided by RefSeq

Specifications

Antigen PPIB
Applications ELISA, Western Blot
Classification Monoclonal
Clone k2E2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant PPIB.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene PPIB
Gene Alias CYP-S1/CYPB/MGC14109/MGC2224/SCYLP
Gene Symbols PPIB
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 26-216 of human PPIB.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5479
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.