Learn More
PPIC, Mouse, Clone: AT3C6, Abnova™
Mouse monoclonal antibody raised against partial recombinant PPIC.
Supplier: Abnova Corporation MAB11113
Description
The protein encoded by this gene is a member of the peptidyl-prolyl cis-trans isomerase (PPIase)) family. PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins. Similar to other PPIases, this protein can bind immunosuppressant cyclosporin A. [provided by RefSeq
Specifications
PPIC | |
Monoclonal | |
1 mg/mL | |
Mouse monoclonal antibody raised against partial recombinant PPIC. | |
In PBS, pH 7.4 (0.09% sodium azide) | |
CYPC/MGC3673 | |
Mouse | |
Protein G purification | |
RUO | |
5480 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2b κ |
ELISA, Western Blot | |
AT3C6 | |
Unconjugated | |
Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
PPIC | |
PPIC | |
Recombinant protein corresponding to amino acids 39-197 of human PPIC. | |
100 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.