Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPM1G, Mouse, Clone: k1G6, Abnova™

Catalog No. 89110787 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-787 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-787 Supplier Abnova Corporation Supplier No. MAB2045
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant PPM1G.

The protein encoded by this gene is a member of the PP2C family of Ser/Thr protein phosphatases. PP2C family members are known to be negative regulators of cell stress response pathways. This phosphatase is found to be responsible for the dephosphorylation of Pre-mRNA splicing factors, which is important for the formation of functional spliceosome. Studies of a similar gene in mice suggested a role of this phosphatase in regulating cell cycle progression. Alternatively spliced transcript variants encoding the same protein have been described. [provided by RefSeq

Specifications

Antigen PPM1G
Applications ELISA, Western Blot
Classification Monoclonal
Clone k1G6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant PPM1G.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (10% glycerol, 0.09% sodium azide)
Gene PPM1G
Gene Alias MGC1675/MGC2870/PP2CG/PP2CGAMMA/PPP2CG
Gene Symbols PPM1G
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 317-546 of human PPM1G.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5496
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.