Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP1R9B, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107462 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-462 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-462 Supplier Abnova Corporation Supplier No. H00084687A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant PPP1R9B.

Spinophilin is a regulatory subunit of protein phosphatase-1 catalytic subunit (PP1; see MIM 176875) and is highly enriched in dendritic spines, specialized protrusions from dendritic shafts that receive most of the excitatory input in the central nervous system (Allen et al., 1997 [PubMed 9275233]).[supplied by OMIM

Sequence: QLEQSVEENKERMEKLEGYWGEAQSLCQAVDEHLRETQAQYQALERKYSKAKRLIKDYQQKEIEFLKKETAQRRVLEESELARKEEMDKLLDKISELEGNLQTLRNSNST

Specifications

Antigen PPP1R9B
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PPP1R9B.
Formulation 50% glycerol
Gene PPP1R9B
Gene Accession No. NM_032595
Gene Alias FLJ30345/PPP1R6/PPP1R9/SPINO/Spn
Gene Symbols PPP1R9B
Host Species Mouse
Immunogen PPP1R9B (NP_115984, 708 a.a. ∽ 817 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84687
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.