Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP2CA Mouse anti-Human, Monoclonal Antibody, Abnova™

Catalog No. 89113544 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-544 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-544 Supplier Abnova Corporation Supplier No. MAB8726
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant PPP2CA.

This gene encodes the phosphatase 2A catalytic subunit. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. This gene encodes an alpha isoform of the catalytic subunit. [provided by RefSeq

Specifications

Antigen PPP2CA
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant PPP2CA.
Dilution Western Blot (1:100-1:2000) ELISA (1:5000-1:20000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.02% sodium azide, 1% BSA, 50% glycerol)
Gene PPP2CA
Gene Accession No. NM_002715
Gene Alias PP2Ac/PP2CA/RP-C
Gene Symbols PPP2CA
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human PPP2CA.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5515
Test Specificity This antibody is specific to PPP2CA.
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.