Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP2R5C, Mouse, Clone: TQ11-1G6, Abnova™

Catalog No. 89110985 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-985 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-985 Supplier Abnova Corporation Supplier No. MAB2390

Mouse monoclonal antibody raised against PPP2R5C.

The product of this gene belongs to the phosphatase 2A regulatory subunit B family. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes a gamma isoform of the regulatory subunit B56 subfamily. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq

Specifications

Antigen PPP2R5C
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone TQ11-1G6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against PPP2R5C.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene PPP2R5C
Gene Alias B56G/MGC23064/PR61G
Gene Symbols PPP2R5C
Host Species Mouse
Immunogen Human PPP2R5C.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5527
Test Specificity This antibody is specific to PP2A (B56 gamma 3 subunit) protein. This antibody does not recognize the B56 gamma 1 isoform of PPP2R5C.
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.