Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prdm1, Mouse, Clone: 3H2-E8, Abnova™

Catalog No. 89110934 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-934 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-934 Supplier Abnova Corporation Supplier No. MAB2327
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant Prdm1.

Specifications

Antigen Prdm1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 3H2-E8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant Prdm1.
Dilution Flow Cytometry (8 ug/mL, 2*10^6/mL cells) Immunocytochemistry (1:50-1:100) Immunofluorescence (1:200) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200) Western Blot (1:1000) The optimal working dilution should be
Formulation In PBS (0.09% sodium azide)
Gene Prdm1
Gene Alias Blimp-1/Blimp1/PRDI-BF1/ZNFPR1A1
Gene Symbols Prdm1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 199-409 of mouse Prdm1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12142
Test Specificity This antibody is specific to the Blimp-1. Recognizes a band ∼98 KDa and may recognize one ∼80 KDa (the beta form).
Target Species Human, Mouse, Pig
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.