Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRDM1 (B01P), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89104152 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-152 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-152 Supplier Abnova Corporation Supplier No. H00000639B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human PRDM1 protein.

This gene encodes a protein that acts as a repressor of beta-interferon gene expression. The protein binds specifically to the PRDI (positive regulatory domain I element) of the beta-IFN gene promoter. Transcription of this gene increases upon virus induction. Two alternatively spliced transcript variants that encode different isoforms have been reported. [provided by RefSeq

Sequence: MKMDMEDADMTLWTEAEFEEKCTYIVNDHPWDSGADGGTSVQAEASLPRNLLFKYATNSEEVIGVMSKEYIPKGTRFGPLIGEIYTNDTVPKNANRKYFWRIYSRGELHHFIDGFNEEKSNWMRYVNPAHSPREQNLAACQNGMNIYFYTIKPIPANQELLVWYCRDFAERLHYPYPGELTMMNLTQTQSSLKQPSTEKNELCPKNVPKREYSVKEILKLDSNPSKGKDLYRSNISPLTSEKDLDDFRRRGSPEMPFYPRVVYPIRAPLPEDFLKASLAYGIERPTYITRSPIPSSTTPSPSARSSPDQSLKSSSPHSSPGNTVSPVGPGSQEHRDSYAYLNASYGTEGLGSYPGYAPLPHLPPAFIPSYNAHYPKFLLPPYGMNCNGLSAVSSMNGINNFGLFPRLCPVYSNLLGGGSLPHPMLNPTSLPSSLPSDGARRLLQPEHPREVLVPAPHSAFSFTGAAASMKDKACSPTSGSPTAGTAATAEHVVQPKATSAAMAAPSSDEAMNLIKNKRNMTGYKTLPYPLKKQNGKIKYECNVCAKTFGQLSNLKVHLRVHSGERPFKCQTCNKGFTQLAHLQKHYLVHTGEKPHECQVCHKRFSSTSNLKTHLRLHSGEKPYQCKVCPAKFTQFVHLKLHKRLHTRERPHKCSQCHKNYIHLCSLKVHLKGNCAAAPAPGLPLEDLTRINEEIEKFDISDNADRLEDVEDDISVISVVEKEILAVVRKEKEETGLKVSLQRNMGNGLLSSGCSLYESSDLPLMKLPPSNPLPLVPVKVKQETVEPMDP

Specifications

Antigen PRDM1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human PRDM1 protein.
Formulation PBS with no preservative; pH 7.4
Gene PRDM1
Gene Accession No. NM_001198.2
Gene Alias BLIMP1/MGC118922/MGC118923/MGC118924/MGC118925/PRDI-BF1
Gene Symbols PRDM1
Host Species Mouse
Immunogen PRDM1 (NP_001189.1,1 a.a. ∽ 789 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 639
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.