Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRDM2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105777 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-777 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-777 Supplier Abnova Corporation Supplier No. H00007799A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant PRDM2.

This tumor suppressor gene is a member of a nuclear histone/protein methyltransferase superfamily. It encodes a zinc finger protein that can bind to retinoblastoma protein, estrogen receptor, and the TPA-responsive element (MTE) of the heme-oxygenase-1 gene. Although the functions of this protein have not been fully characterized, it may (1) play a role in transcriptional regulation during neuronal differentiation and pathogenesis of retinoblastoma, (2) act as a transcriptional activator of the heme-oxygenase-1 gene, and (3) be a specific effector of estrogen action. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: KTSRSLHVRVQKSKAVLQSKSTLASKKRTDRFNIKSRERSGGPVTRSLQLAAAADLSENKREDGSAKQELKDFSYSLRLASRCSPPAAPYITRQYRKVKA

Specifications

Antigen PRDM2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PRDM2.
Formulation 50% glycerol
Gene PRDM2
Gene Accession No. NM_012231
Gene Alias HUMHOXY1/KMT8/MTB-ZF/RIZ/RIZ1/RIZ2
Gene Symbols PRDM2
Host Species Mouse
Immunogen PRDM2 (NP_036363, 1606 a.a. ∽ 1705 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7799
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.