Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-PRDM5, Clone: 57-20,

Catalog No. MABE972MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE972MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE972MI Supplier MilliporeSigma™ Supplier No. MABE972
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects PRDM5 clone: 57-20 in Mouse samples, and it is validated for Immunoprecipitation, Western Blotting

PR domain zinc finger protein 5 (UniProt Q9NQX1) is encoded by the PRDM1 gene (also known as BCS2, PFM2) in human (Gene ID 11107). PRDM proteins constitute a family of transcriptional regulators that are characterized by the presence of variable numbers of zinc finger domains and an N-terminal PR domain. PRDM5 can negatively regulate gene transcriptions by recruiting the G9a histone methyltransferase and histone deacetylases. PRDM5 can promote transcription by targeting enhancer-like elements or recruiting RNA polymerase II binding. PRDM5 is highly expressed in murine embryonic stem cells (mESCs) and associates with complexes involved in chromatin organization, such as Ctcf, Smc1 (cohesin), and TFIIIC, which indicates their involvement in chromatin organization. PRDM5 is often found silenced in human breast, ovarian, liver, and cervical cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDM5
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 57-20
Formulation Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q9NQX1
Gene Symbols PRDM5; PFM2
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human PRDM5 near the N-terminus.
Purification Method Protein G Purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_061169
Target Species Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.