Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRDX6, Mouse, Clone: 5, Abnova™

Catalog No. 89113865 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-865 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-865 Supplier Abnova Corporation Supplier No. MAB9907
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant PRDX6.

The protein encoded by this gene is a member of the thiol-specific antioxidant protein family. This protein is a bifunctional enzyme with two distinct active sites. It is involved in redox regulation of the cell; it can reduce H(2)O(2) and short chain organic, fatty acid, and phospholipid hydroperoxides. It may play a role in the regulation of phospholipid turnover as well as in protection against oxidative injury. [provided by RefSeq

Specifications

Antigen PRDX6
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant PRDX6.
Dilution Western blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300)
Gene PRDX6
Gene Accession No. NM_004905.2
Gene Alias 1-Cys/AOP2/KIAA0106/MGC46173/NSGPx/PRX/aiPLA2/p29
Gene Symbols PRDX6
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human PRDX6.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9588
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.