Learn More
PRELP, Mouse, Clone: 3H1, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PRELP.
Supplier: Abnova Corporation H00005549M01A
Description
The protein encoded by this gene is a leucine-rich repeat protein present in connective tissue extracellular matrix. This protein functions as a molecule anchoring basement membranes to the underlying connective tissue. This protein has been shown to bind type I collagen to basement membranes and type II collagen to cartilage. It also binds the basement membrane heparan sulfate proteoglycan perlecan. This protein is suggested to be involved in the pathogenesis of Hutchinson-Gilford progeria (HGP), which is reported to lack the binding of collagen in basement membranes and cartilage. Alternatively spliced transcript variants encoding the same protein have been observed. [provided by RefSeq
Sequence: RGLPKNSFNISNLLVLHLSHNRISSVPAINNRLEHLYLNNNSIEKINGTQICPNDLVAFHDFSSDLENVPHLRYLRLDGNYLKPPSpecifications
PRELP | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
NM_002725 | |
PRELP | |
PRELP (NP_002716.1,281 a.a. ∽ 365 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
5549 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
3H1 | |
Mouse monoclonal antibody raised against a partial recombinant PRELP. | |
PRELP | |
MGC45323/MST161/MSTP161/SLRR2A | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgM κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.