Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prf1, Mouse, Clone: CE2.10, Abnova™

Catalog No. 89111553 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-553 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-553 Supplier Abnova Corporation Supplier No. MAB3711
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant Prf1.

Specifications

Antigen Prf1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone CE2.10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant Prf1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene Prf1
Gene Alias Pfn/Pfp/Prf-1
Gene Symbols Prf1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 98-534 of mouse Prf1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18646
Test Specificity Recognizes mouse and human perforin.
Target Species Human, Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.