Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRICKLE1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107637 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-637 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-637 Supplier Abnova Corporation Supplier No. H00144165A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant PRICKLE1.

This gene encodes a nuclear receptor that may be a negative regulator of the Wnt/beta-catenin signaling pathway. The encoded protein localizes to the nuclear membrane and has been implicated in the nuclear trafficking of the transcription repressors REST/NRSF and REST4. Mutations in this gene have been linked to progressive myoclonus epilepsy. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 3. [provided by RefSeq

Sequence: DNALNLVTERKYSPKDRLRLYTPDNYEKFIQNKSAREIQAYIQNADLYGQYAHATPDYGLQNPGMNRFLGLYG

Specifications

Antigen PRICKLE1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PRICKLE1.
Formulation 50% glycerol
Gene PRICKLE1
Gene Accession No. NM_153026
Gene Alias EPM1B/FLJ31627/FLJ31937/MGC138902/MGC138903/RILP
Gene Symbols PRICKLE1
Host Species Mouse
Immunogen PRICKLE1 (NP_694571, 684 a.a. ∽ 756 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144165
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.