Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKACA, Mouse, Clone: 3C1, Abnova™

Catalog No. 89111155 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-155 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-155 Supplier Abnova Corporation Supplier No. MAB2905

Mouse monoclonal antibody raised against native PRKACA.

Specifications

Antigen PRKACA
Applications ELISA
Classification Monoclonal
Clone 3C1
Conjugate Unconjugated
Description protein kinase, cAMP-dependent, catalytic, alpha
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene PRKACA
Gene Symbols PRKACA
Host Species Mouse
Immunogen Native purified bovine cardiac muscle PRKACA.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282322
Target Species Bovine
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.