Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKACA, Mouse, Clone: 6D2.1, Abnova™

Catalog No. 89111491 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-491 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-491 Supplier Abnova Corporation Supplier No. MAB3591
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of PRKACA.

cAMP is a signaling molecule important for a variety of cellular functions. cAMP exerts its effects by activating the cAMP-dependent protein kinase, which transduces the signal through phosphorylation of different target proteins. The inactive kinase holoenzyme is a tetramer composed of two regulatory and two catalytic subunits. cAMP causes the dissociation of the inactive holoenzyme into a dimer of regulatory subunits bound to four cAMP and two free monomeric catalytic subunits. Four different regulatory subunits and three catalytic subunits have been identified in humans. The protein encoded by this gene is a member of the Ser/Thr protein kinase family and is a catalytic subunit of cAMP-dependent protein kinase. Alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq

Sequence: ESPAQNTAHLDQFERIK

Specifications

Antigen PRKACA
Applications Dot Blot, Western Blot
Classification Monoclonal
Clone 6D2.1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of PRKACA.
Dilution Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene PRKACA
Gene Alias MGC102831/MGC48865/PKACA
Gene Symbols PRKACA
Host Species Mouse
Immunogen A synthetic peptide corresponding to human PRKACA.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5566
Test Specificity This antibody strongly reacts with human proteinkinase A catalytic (PRKACA) alpha subunit, and weakly with PKAc gamma subunit (both around 40 KDa). The recognized epitope of PKAc alpha is identical between man, sheep, pig, ox and dog.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.