Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKAR2B, Mouse, Clone: RS17, Abnova™

Catalog No. 89108010 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-010 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-010 Supplier Abnova Corporation Supplier No. MAB0621

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse monoclonal antibody raised against native bovine PRKAR2B.

Specifications

Antigen PRKAR2B
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone RS17
Conjugate Unconjugated
Description protein kinase, cAMP-dependent, regulatory, type II, beta
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene PRKAR2B
Gene Symbols PRKAR2B
Host Species Mouse
Immunogen Native purified bovine PRKAR2B.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282463
Target Species Bovine
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.