Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prkcq, Mouse, Clone: M217, Abnova™

Catalog No. 89110404 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-404 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-404 Supplier Abnova Corporation Supplier No. MAB1389
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant Prkcq.

Specifications

Antigen Prkcq
Applications ELISA, Western Blot
Classification Monoclonal
Clone M217
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant Prkcq.
Dilution ELISA (1:2000) Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide)
Gene Prkcq
Gene Alias A130035A12Rik/AW494342/PKC-0/PKC-theta/PKCtheta/Pkcq
Gene Symbols Prkcq
Host Species Mouse
Immunogen Recombinant protein corresponding to N-terminus of mouse Prkcq.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18761
Test Specificity This sequence is conserved in human and rat PKCq, and has low homology to other PKC family members.
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.