Learn More
PRKD2, Mouse, Clone: 2E4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PRKD2.
Supplier: Abnova Corporation H00025865M01A
Description
The protein encoded by this gene belongs to the protein kinase D (PKD) family of serine/threonine protein kinases. This kinase can be activated by phorbol esters as well as by gastrin via the cholecystokinin B receptor (CCKBR) in gastric cancer cells. It can bind to diacylglycerol (DAG) in the trans-Golgi network (TGN) and may regulate basolateral membrane protein exit from TGN. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq
Sequence: MDESEDSGVIPGSHSENALHASEEEEGEGGKAQSSLGYIPLMRVVQSVRHTTRKSSTTLREGWVVHYSNKDTLRKRHYWRLDCKCITLFQNNTTNRYYKEIPLSEILTVESpecifications
PRKD2 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
BC025307.1 | |
PRKD2 | |
PRKD2 (AAH25307.1,1 a.a. ∽ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
25865 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
2E4 | |
Mouse monoclonal antibody raised against a partial recombinant PRKD2. | |
PRKD2 | |
HSPC187/PKD2 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.