Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKDC, Mouse, Clone: 18-2, 25-4, 42-psc, Abnova™

Catalog No. 89111416 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-416 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-416 Supplier Abnova Corporation Supplier No. MAB3433

Mouse monoclonal antibody raised against native PRKDC.

The PRKDC gene encodes the catalytic subunit of a nuclear DNA-dependent serine/threonine protein kinase (DNA-PK). The second component is the autoimmune antigen Ku (MIM 152690), which is encoded by the G22P1 gene on chromosome 22q. On its own, the catalytic subunit of DNA-PK is inactive and relies on the G22P1 component to direct it to the DNA and trigger its kinase activity; PRKDC must be bound to DNA to express its catalytic properties.[supplied by OMIM

Specifications

Antigen PRKDC
Applications Immunofluorescence, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 18-2, 25-4, 42-psc
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native PRKDC.
Dilution Immunoprecipitation (2 ug/mg protein lysate) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.4 (0.09% sodium azide)
Gene PRKDC
Gene Alias DNA-PKcs/DNAPK/DNPK1/HYRC/HYRC1/XRCC7/p350
Gene Symbols PRKDC
Host Species Mouse
Immunogen Native purified PRKDC from HeLa cells.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5591
Test Specificity Reacts with the nuclear catalytic subunit of DNA-dependent Protein Kinase (DNA-PKcs). Epitopes of the three clones have not been precisely mapped, but experimental data suggests that they are different.
Target Species Chicken, Frog, Hamster, Human, Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1, IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.