Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRL, Mouse, Clone: 090-10145, Abnova™
Mouse monoclonal antibody raised against native PRL.
Supplier: Abnova Corporation MAB4299
Specifications
PRL | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
PRL | |
Mouse | |
1 mg | |
Primary | |
Human | |
Liquid |
ELISA | |
090-10145 | |
Mouse monoclonal antibody raised against native PRL. | |
In 10mM phosphate buffer, pH 7.4 (150mM NaCl, 0.09% sodium azide) | |
PRL | |
Native purified human PRL. | |
RUO | |
5617 | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction