Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRL, Mouse, Clone: 090-11710, Abnova™

Catalog No. 89111757 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-757 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-757 Supplier Abnova Corporation Supplier No. MAB4300
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against PRL.

Specifications

Antigen PRL
Applications ELISA
Classification Monoclonal
Clone 090-11710
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against PRL.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM phosphate buffer, pH 7.4 (150mM NaCl, 0.09% sodium azide)
Gene PRL
Gene Symbols PRL
Host Species Mouse
Immunogen Human Pituitary Gland.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5617
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.