Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRNP, Mouse, Clone: EM-20, Abnova™

Catalog No. 89112887 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-887 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-887 Supplier Abnova Corporation Supplier No. MAB6472
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant PRNP.

The protein encoded by this gene is a membrane glycosylphosphatidylinositol-anchored glycoprotein that tends to aggregate into rod-like structures. The encoded protein contains a highly unstable region of five tandem octapeptide repeats. This gene is found on chromosome 20, approximately 20kb upstream of a gene which encodes a biochemically and structurally similar protein to the one encoded by this gene. Mutations in the repeat region as well as elsewhere in this gene have been associated with Creutzfeldt-Jakob disease, fatal familial insomnia, Gerstmann-Straussler disease, Huntington disease-like 1, and kuru. Alternative splicing results in multiple transcript variants encoding the same protein. [provided by RefSeq

Specifications

Antigen PRNP
Applications Western Blot
Classification Monoclonal
Clone EM-20
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant PRNP.
Dilution Western Blot (0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene PRNP
Gene Alias ASCR/CD230/CJD/GSS/MGC26679/PRIP/PrP/PrP27-30/PrP33-35C/PrPc/prion
Gene Symbols PRNP
Host Species Mouse
Immunogen Recombinant protein corresponding to human PRNP.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5621
Test Specificity This antibody recognizes human prion protein (PrP). Diglycosylated form of PrP has ∼40 KDa, monoglycosylated form ∼30 KDa, and nonglycosylated form ∼19-21 KDa. This antibody is suitable for discrimination between normal cellular prion protein (PrPc) and its conformationally changed form (PrPSc) prionprotein.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.