Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Progesterone, Mouse, Clone: XM207, Abnova™

Catalog No. 89135275 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-275 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-275 Supplier Abnova Corporation Supplier No. MAB2858
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Progesterone.

Specifications

Antigen Progesterone
Applications ELISA
Classification Monoclonal
Clone XM207
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Progesterone.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM Tris, 150mM NaCl, pH 7.5 (0.05% sodium azide)
Host Species Mouse
Immunogen Progesterone conjugated with BSA.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity There is no cross-reactivity with BSA.
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.