Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prolactin, Mouse, Clone: 6F11, Abnova™

Catalog No. 89110903 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-903 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-903 Supplier Abnova Corporation Supplier No. MAB2284
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Prolactin.

Specifications

Antigen Prolactin
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 6F11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Prolactin.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Gene Symbols PRL
Host Species Mouse
Immunogen A mixture native PRL of sheep and human.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5617/443317
Test Specificity The epitope recognized by this antibody is restricted to a region that includes the carboxyl-terminal disulfide loop that is conserved among prolactins from several species.
Target Species Bovine, Human, Monkey, Mouse, Rat, Sheep
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.