Learn More
PRPF3, Mouse, Clone: 3H6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PRPF3.
Supplier: Abnova Corporation H00009129M01A
Description
The removal of introns from nuclear pre-mRNAs occurs on complexes called spliceosomes, which are made up of 4 small nuclear ribonucleoprotein (snRNP) particles and an undefined number of transiently associated splicing factors. This gene product is one of several proteins that associate with U4 and U6 snRNPs. Mutations in this gene are associated with retinitis pigmentosa-18. [provided by RefSeq
Sequence: EGGPKAQKKFKRLMLHRIKWDEQTSNTKGDDDEESDEEAVKKTNKCVLVWEGTAKDRSFGEMKFKQCPTENMAREHFKKHGAEHYWDLALSESVLESTDSpecifications
PRPF3 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
NM_004698 | |
PRPF3 | |
PRPF3 (NP_004689,585 a.a. ∽ 683 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
9129 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
3H6 | |
Mouse monoclonal antibody raised against a partial recombinant PRPF3. | |
PRPF3 | |
HPRP3/HPRP3P/PRP3/Prp3p/RP18 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG3 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.