Learn More
PRPS1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant PRPS1.
Supplier: Abnova Corporation H00005631A01
Description
This gene encodes an enzyme that catalyzes the phosphoribosylation of ribose 5-phosphate to 5-phosphoribosyl-1-pyrophosphate, which is necessary for purine metabolism and nucleotide biosynthesis. Defects in this gene are a cause of phosphoribosylpyrophosphate synthetase superactivity, Charcot-Marie-Tooth disease X-linked recessive type 5 and Arts Syndrome. [provided by RefSeq
Sequence: MPNIKIFSGSSHQDLSQKIADRLGLELGKVVTKKFSNQETCVEIGESVRGEDVYIVQSGCGEINDNLMELLIMINACKIASASRVTAVIPCFPYARQDKKSpecifications
| PRPS1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant PRPS1. | |
| PRPS1 | |
| ARTS/CMTX5/KIAA0967/PPRibP/PRSI | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_002764 | |
| PRPS1 | |
| PRPS1 (NP_002755, 1 a.a. ∽ 100 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 5631 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only