Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRPS1L1, Mouse, Clone: 3, Abnova™

Catalog No. 89113886 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-886 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-886 Supplier Abnova Corporation Supplier No. MAB9928
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant PRPS1L1.

This intronless gene is specifically expressed in the testis, and encodes a protein that is highly homologous to the two subunits of phosphoribosylpyrophosphate synthetase encoded by human X-linked genes, PRPS1 and PRPS2. These enzymes convert pyrimidine, purine or pyridine bases to the corresponding ribonucleoside monophosphates. In vitro transcription/translation and site-directed mutagenesis studies indicate that translation of this mRNA initiates exclusively at a non-AUG (ACG) codon. [provided by RefSeq

Specifications

Antigen PRPS1L1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant PRPS1L1.
Dilution Western blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300)
Gene PRPS1L1
Gene Accession No. NM_175886
Gene Alias PRPS1/PRPS3/PRPSL/PRS-III
Gene Symbols PRPS1L1
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human PRPS1L1.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221823
Test Specificity It can expression in SW480 whole cell lysate.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.