Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRR7, Mouse, Clone: TRAP3/10, Abnova™

Catalog No. 89113002 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-002 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-002 Supplier Abnova Corporation Supplier No. MAB6689
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant PRR7.

Specifications

Antigen PRR7
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone TRAP3/10
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant PRR7.
Dilution Immunocytochemistry (10 ug/mL) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene PRR7
Gene Alias MGC10772
Gene Symbols PRR7
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 126-253 of human PRR7.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80758
Test Specificity This antibody recognizes PRR7, a 28 KDa proline-rich membrane protein presumably associated with NMDA receptor complex.
Target Species Human, Mouse
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.