Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRTN3, Mouse, Clone: PR3-G2, Abnova™

Catalog No. 89112161 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-161 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-161 Supplier Abnova Corporation Supplier No. MAB5194
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native PRTN3.

Specifications

Antigen PRTN3
Applications ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone PR3-G2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native PRTN3.
Dilution Recommend dilution at 1:50 The optimal working dilution should be determined by the end user.
Formulation In PBS (0.1% BSA, 0.02% sodium azide)
Gene PRTN3
Gene Alias ACPA/AGP7/C-ANCA/MBT/P29/PR-3
Gene Symbols PRTN3
Host Species Mouse
Immunogen Native purified human PRTN3.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5657
Test Specificity Monoclonal antibody PR3G-2 reacts with human proteinase 3 (PR3), a 30 KDa protein. The monoclonal antibody PR3G-2 does not inhibit the biological activity of PR-3.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.