Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Psd, Mouse, Clone: 7E3, Abnova™

Catalog No. 89112959 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-959 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-959 Supplier Abnova Corporation Supplier No. MAB6625
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant Psd.

Specifications

Antigen Psd
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 7E3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant Psd.
Dilution Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Psd
Gene Alias EFA6
Gene Symbols Psd
Host Species Mouse
Immunogen Recombinant protein corresponding to rat Psd.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171381
Test Specificity Detects a ∼100KDa protein corresponding to the molecular mass of PSD-95 on SDS PAGE immunoblots. Additional cross-reactive bands are detected at ∼80KDa and ∼50KDa in rat and mouse samples.
Target Species Bovine, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.