Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSEN1, Mouse, Clone: APS11, Abnova™

Catalog No. 89112355 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-355 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-355 Supplier Abnova Corporation Supplier No. MAB5677

Mouse monoclonal antibody raised against synthetic peptide of PSEN1.

Alzheimer's disease (AD) patients with an inherited form of the disease carry mutations in the presenilin proteins (PSEN1; PSEN2) or in the amyloid precursor protein (APP). These disease-linked mutations result in increased production of the longer form of amyloid-beta (main component of amyloid deposits found in AD brains). Presenilins are postulated to regulate APP processing through their effects on gamma-secretase, an enzyme that cleaves APP. Also, it is thought that the presenilins are involved in the cleavage of the Notch receptor, such that they either directly regulate gamma-secretase activity or themselves are protease enzymes. Several alternatively spliced transcript variants encoding different isoforms have been identified for this gene, the full-length nature of only some have been determined. [provided by RefSeq]

Sequence: HLSNTVRSQNDNRE

Specifications

Antigen PSEN1
Applications ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone APS11
Conjugate Unconjugated
Description presenilin 1
Formulation Lyophilized from PBS
Gene PSEN1
Gene Alias AD3, FAD, PS1, S182
Gene Symbols PSEN1
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 21-34 of human PSEN1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5663
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C or -20°C on dry atmosphere. After reconstitution with water and glycerol (1:1), store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.