Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMA8, Mouse, Clone: 5A8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PSMA8.
Supplier: Abnova Corporation H00143471M04A
Description
Sequence: GFDDDGISRLYQTDPSGTYHAWKANAIGRSAKTVREFLEKNYTEDAIASDSEAIKLAIKALLEVVQSGGRNIELAIIRRNQPLKMFSAKEVELYVTEIEKSpecifications
PSMA8 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
NM_144662 | |
PSMA8 | |
PSMA8 (NP_653263,143 a.a. ∽ 242 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
143471 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
5A8 | |
Mouse monoclonal antibody raised against a partial recombinant PSMA8. | |
PSMA8 | |
MGC26605/PSMA7L | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction