Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Psmd4, Mouse, Clone: AH1.1, Abnova™

Catalog No. 89113160 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-160 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-160 Supplier Abnova Corporation Supplier No. MAB7301

Mouse monoclonal antibody raised against Psmd4.

Specifications

Antigen Psmd4
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone AH1.1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Psmd4.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene Psmd4
Gene Alias Af1/Mcb1/angiocidin
Gene Symbols Psmd4
Host Species Mouse
Immunogen Mouse Psmd4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19185
Test Specificity Proteasome S5A (AH1.1). Not yet tested in other species. Predicted molecular weight: 41 KDa. Not tested in other applications.
Target Species Mouse
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.