Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSPH, Mouse, Clone: 3A5, Abnova™

Catalog No. 89105243 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-243 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-243 Supplier Abnova Corporation Supplier No. H00005723M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant PSPH.

The protein encoded by this gene belongs to a subfamily of the phosphotransferases. This encoded enzyme is responsible for the third and last step in L-serine formation. It catalyzes magnesium-dependent hydrolysis of L-phosphoserine and is also involved in an exchange reaction between L-serine and L-phosphoserine. Deficiency of this protein is thought to be linked to Williams syndrome. [provided by RefSeq

Sequence: MVSHSELRKLFYSADAVCFDVDSTVIREEGIDELAKICGVEDAVSEMTRRAMGGAVPFKAALTERLALIQPSREQVQRLIAEQPPHLTPGIRELVSRLQERNVQVFLISGGFRSIVEHVASKLNIPATNVFANRLKFYFNGEYAGFDETQPTAESGGKGKVIKLLKEKFHFKKIIMIGDGATDMEACPPADAFIGFGGNVIRQQVKDNAKWYITDFVELLGELEE

Specifications

Antigen PSPH
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3A5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant PSPH.
Formulation ascites with no preservative
Gene PSPH
Gene Accession No. BC063614
Gene Alias PSP
Gene Symbols PSPH
Host Species Mouse
Immunogen PSPH (AAH63614,1 a.a. ∽ 225 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5723
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.