Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Mouse anti-PTB, Clone: Bb7,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABE986
Description
Specifically detects PTB clone: Bb7 in Human, Mouse samples, and it is validated for Immunocytochemistry, Immunoprecipitation, Western Blotting
Polypyrimidine tract-binding protein 1 (UniProt P26599; also known as 57kDa RNA-binding protein PPTB-1, Heterogeneous nuclear ribonucleoprotein I, Heterogeneous nuclear ribonucleoprotein polypeptide I, hnRNP I, PTB, RNA-binding protein) is encoded by the PTBP1 (also known as HNRNP-I, HNRNPI, HNRPI, PTB, PTB2, PTB3, PTB4, PTB-1, PTB-T) gene (Gene ID 5725) in human. The serine–arginine rich (SR) proteins and the heterogeneous nuclear ribonucleoproteins (hnRNPs) are the most typical families of RBPs involved in splicing regulation. SR proteins generally play a positive role in exon recognition by binding exonic and intronic enhancers near the alternative splice sites. On the other hand, hnRNPs often bind to silencer elements and interfere with the recruitment of spliceosome components or positive regulators. PTB, also known as hnRNP I, belongs to an hnRNP family of RNA-binding proteins and is involved in several aspects of cellular mRNA metabolism including splicing, RNA stability, and internal ribosome entry site (IRES)-mediated translation of viral and cellular mRNAs. PTB functions as a splicing repressor by preventing recruitment of U2AF65 or correct base pairing of U2 snRNP, or by affecting the interaction of U1 snRNP with other components of the spliceosome.
Specifications
PTB | |
Monoclonal | |
Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
PTBP1; PTB | |
Recombinant protein human PTB. | |
100 μg | |
Epigenetics and Nuclear Function | |
NP_002810 | |
Stable for one year at 2°C to 8°C from date of receipt. | |
IgG2b κ |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
Bb7 | |
P26599 | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction