Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTBP2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107222 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-222 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-222 Supplier Abnova Corporation Supplier No. H00058155A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length recombinant PTBP2.

The protein encoded by this gene binds to the intronic cluster of RNA regulatory elements, downstream control sequence (DCS). It is implicated in controlling the assembly of other splicing-regulatory proteins. This protein is very similar to the polypyrimidine tract binding protein but it is expressed primarily in the brain. [provided by RefSeq

Sequence: MDGIVTEVAVGVKRGSDELLSGSVLSSPNSNMSSMVVTANGNDSKKFKGEDKMDGAPSRVLHIRKLPGEVTETEVIALGLPFGKVTNILMLKGKNQAFLELATEEAAITMVNYYSAVTPHLRNQPIYIQYSNHKELKTDNTLNQRAQAVLQAVTAVQTANTPLSGTTVSESAVTPAQSPVLRIIIDNMYYPVTLDVLHQIFSKFGAVLKIITFTKNNQFQALLQYGDPVNAQQAKLALDGQNIYNACCTLRIDFSKLVNLNVKYNNDKSRDYTRPDLPSGDGQPALDPAIAAAFAKETSLLAVPGALSPLAIPNAAAAAAAAAAGRVGMPGVSAGGNTVLLVSNLNEEMVTPQSLFTLFGVYGDVQRVKILYNKKDSALIQMADGNQSQLAMNHLNGQKMYGKIIRVTLSKHQTVQLPREGLDDQGLTKDFGNSPLHRFKKPGSKNFQNIFPPSATLHLSNIPPSVAEEDLRTLFANTGGTVKAFKFFQRDHKMALLQMATVEEAIQALIDLHNYNLGENHHLRVSFSKSTI

Specifications

Antigen PTBP2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant PTBP2.
Formulation 50% glycerol
Gene PTBP2
Gene Accession No. BC016582
Gene Alias FLJ34897/PTB/PTBLP/brPTB/nPTB/nPTB5/nPTB6/nPTB7/nPTB8
Gene Symbols PTBP2
Host Species Mouse
Immunogen PTBP2 (AAH16582, 1 a.a. ∽ 532 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58155
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.