Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTGER2, Mouse, Clone: 3E6, Abnova™

Catalog No. 89110741 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-741 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-741 Supplier Abnova Corporation Supplier No. MAB1976
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant PTGER2.

Specifications

Antigen PTGER2
Applications Western Blot
Classification Monoclonal
Clone 3E6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant PTGER2.
Dilution Western blot (1 to 5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Gene PTGER2
Gene Alias EP2
Gene Symbols PTGER2
Host Species Mouse
Immunogen Recombinant protein of human PTGER2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5732
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.