Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTGER3, Mouse, Clone: 5F5, Abnova™

Catalog No. 89110743 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-743 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-743 Supplier Abnova Corporation Supplier No. MAB1978
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant PTGER3.

The protein encoded by this gene is a member of the G-protein coupled receptor family. This protein is one of four receptors identified for prostaglandin E2 (PGE2). This receptor may have many biological functions, which involve digestion, nervous system, kidney reabsorption, and uterine contraction activities. Studies of the mouse counterpart suggest that this receptor may also mediate adrenocorticotropic hormone response as well as fever generation in response to exogenous and endogenous stimuli. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen PTGER3
Applications Western Blot
Classification Monoclonal
Clone 5F5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant PTGER3.
Dilution Western blot (1 to 5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Gene PTGER3
Gene Alias EP3/EP3-I/EP3-II/EP3-III/EP3-IV/EP3e/MGC141828/MGC141829/MGC27302
Gene Symbols PTGER3
Host Species Mouse
Immunogen Recombinant protein of human PTGER3.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5733
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.