Learn More
PTGES, Mouse, Clone: 2B9, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant PTGES.
Supplier: Abnova Corporation H00009536M04A
Description
The protein encoded by this gene is a glutathione-dependent prostaglandin E synthase. The expression of this gene has been shown to be induced by proinflammatory cytokine interleukin 1 beta (IL1B). Its expression can also be induced by tumor suppressor protein TP53, and may be involved in TP53 induced apoptosis. Knockout studies in mice suggest that this gene may contribute to the pathogenesis of collagen-induced arthritis and mediate acute pain during inflammatory responses. [provided by RefSeq
Sequence: MPAHSLVMSSPALPAFLLCSTLLVIKMYVVAIITGQVRLRKKAFANPEDALRHGGPQYCRSDPDVERCLRAHRNDMETIYPFLFLGFVYSFLGPNPFVAWMHFLVFLVGRVAHTVAYLGKLRAPIRSVTYTLAQLPCASMALQILWEAARHLSpecifications
PTGES | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
BC008280 | |
PTGES | |
PTGES (AAH08280.1,1 a.a. ∽ 152 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
9536 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA | |
2B9 | |
Mouse monoclonal antibody raised against a full-length recombinant PTGES. | |
PTGES | |
MGC10317/MGST-IV/MGST1-L1/MGST1L1/MPGES/PGES/PIG12/PP102/PP1294/TP53I12/mPGES-1 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.