Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTGIS, Mouse, Monoclonal Antibody, Abnova™

Catalog No. 89111598 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-598 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-598 Supplier Abnova Corporation Supplier No. MAB3798
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against PTGIS.

Specifications

Antigen PTGIS
Applications ELISA, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against PTGIS.
Dilution ELISA (1:10) Immunoprecipitation (1:2) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.09% sodium azide
Gene PTGIS
Gene Alias MGC140775
Gene Symbols PTGIS
Host Species Mouse
Immunogen Bovine PTGIS.
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282021
Test Specificity Bovine PGI2 Synthase reacts to most mammalian species, including bovine, mouse, rat, sheep, guinea pig, and rabbit.
Target Species Bovine, Guinea Pig, Mouse, Rabbit, Rat, Sheep
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.