Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTGS1 Mouse anti-Bovine, Guinea pig, Human, Rabbit, Sheep, Monoclonal Antibody, Abnova™

Catalog No. 89135514 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-514 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-514 Supplier Abnova Corporation Supplier No. MAB3797
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against PTGS1.

Specifications

Antigen PTGS1
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against PTGS1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.09% sodium azide
Host Species Mouse
Immunogen Sheep PTGS1.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts with sheep, bovine, guinea pig, rabbit and human PGH Synthase. Does not react with rat, mouse or dog PGH Synthase. specific to COX-1 without cross reactivity to Cyclooxygenase 2 (COX-2).
Target Species Bovine, Guinea Pig, Human, Rabbit, Sheep
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.