Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTH, Mouse, Clone: 14H5 1C7, Abnova™

Catalog No. 89111537 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-537 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-537 Supplier Abnova Corporation Supplier No. MAB3690
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant PTH.

The protein encoded by this gene is a hormone secreted by parathyroid cells. This hormone elevates blood Ca2+ level by dissolving the salts in bone and preventing their renal excretion. Defects in this gene are a cause of familial isolated hypoparathyroidism (FIH). [provided by RefSeq]

Specifications

Antigen PTH
Applications Radioimmune Assays (RIA)
Classification Monoclonal
Clone 14H5 1C7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant PTH.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS or in BBS
Gene PTH
Gene Alias PTH1
Gene Symbols PTH
Host Species Mouse
Immunogen Recombinant protein corresponding to human PTH.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5741
Test Specificity This antibody recognizes natural and recombinant human PTH.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.