Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Mouse anti-PTHrP (1-34), Clone: 23-57-137-1,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABN793
Description
Specifically detects PTHrP (1-34) clone: 23-57-137-1 in Human samples, and it is validated for Neutralization, Western Blotting
Parathyroid hormone-related protein (UniProt P12272), also known as Parathyroid hormone-like related protein, Parathyroid hormone-like hormone preproprotein, PLP, PTH-related protein, PTH-rP, PTHrP) is encoded by the PTHLH (also known as BDE2, HHM, PTHR, PTHRP) gene (Entrez Gene ID 5744) in human. Parathyroid hormone-related protein is initially translated as a 177 amino acid protein, which is then cleaved into PTHrP(1-34), PTHrP(1-36), PTHrP(38-94), and Osteostatin (also known as PTHrP (107-139)). Malignancy-associated hypercalcemia is caused by excessive production of parathyroid hormone-related protein (PTHrP) by the tumor. Neutralizing PTHrP(1-34) by antibody injection is shown to reduce serum calcium levels in hypercalcemic nude mice bearing transplanted human PTHrP-producing tumors, leading to their prolonged survival. (PMID 8352067, 11668474, 15517871, 16036863, 19342773)
Specifications
PTHrP (1-34) | |
Monoclonal | |
Purified mouse monoclonal IgG1κ in buffer containing PBS without preservatives. | |
PTHLH; PTHRP | |
Bovine thyroglobulin-conjugated linear peptide corresponding to human PTHrP(1-34). | |
100 μg | |
Neuroscience | |
NP_002811 | |
Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance. | |
IgG1 κ |
Neutralization, Western Blot | |
23-57-137-1 | |
P12272 | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction