Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPN13, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105255 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-255 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-255 Supplier Abnova Corporation Supplier No. H00005783A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant PTPN13.

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP is a large intracellular protein. It has a catalytic PTP domain at its C-terminus and two major structural domains: a region with five PDZ domains and a FERM domain that binds to plasma membrane and cytoskeletal elements. This PTP was found to interact with, and dephosphorylate, Fas receptor and IkappaBalpha through the PDZ domains. This suggests it has a role in Fas mediated programmed cell death. This PTP was also shown to interact with GTPase-activating protein, and thus may function as a regulator of Rho signaling pathways. Four alternatively spliced transcript variants, which encode distinct proteins, have been reported. [provided by RefSeq

Sequence: FTDENISNQDLRAFTAPEVLQNQSLTSLSDVEKIHIYSLGMTLYWGADYEVPQSQPIKLGDHLNSILLGMCEDVIYARVSVRTVLDACSAHIRNSNCAPSFSYVKHLVKL

Specifications

Antigen PTPN13
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PTPN13.
Formulation 50% glycerol
Gene PTPN13
Gene Accession No. NM_006264
Gene Alias DKFZp686J1497/FAP-1/PNP1/PTP-BAS/PTP-BL/PTP1E/PTPL1/PTPLE
Gene Symbols PTPN13
Host Species Mouse
Immunogen PTPN13 (NP_006255, 66 a.a. ∽ 175 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5783
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.