Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ptpn5, Mouse, Clone: 23E5, Abnova™

Catalog No. 89110993 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-993 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-993 Supplier Abnova Corporation Supplier No. MAB2403
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of Ptpn5.

Sequence: MCTPGCNEEGFGYLVSPR

Specifications

Antigen Ptpn5
Applications Western Blot
Classification Monoclonal
Clone 23E5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of Ptpn5.
Dilution Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.09% sodium azide
Gene Ptpn5
Gene Alias STEP
Gene Symbols Ptpn5
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 57-74 of mouse Ptpn5.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29644
Test Specificity This antibody is specific to STEP61, STEP46, STEP38, and STEP20.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.