Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ptprb, Mouse, Clone: 122.2, Abnova™
Mouse monoclonal antibody raised against native Ptprb.
Supplier: Abnova Corporation MAB2429
Specifications
Ptprb | |
Monoclonal | |
Unconjugated | |
Western Blot (1:1000-1:2500) The optimal working dilution should be determined by the end user. | |
Ptprb | |
Mouse | |
100 μL | |
Primary | |
Receptor Tyrosine Phosphatase Beta (phosphacan). | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgM |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
122.2 | |
Mouse monoclonal antibody raised against native Ptprb. | |
In PBS | |
Ptprb | |
Native purified rat Ptprb. | |
RUO | |
314843 | |
Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction